siRNA Details
Gene Symbol: Kdm7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033430.4 | NP_001028602.2 | 20 | 8844 | ||
GenBank mRNA | AK133429.1 | - | 1 | 608 | ||
AK158533.1 | - | 1 | 1861 | |||
AK163747.1 | - | 1 | 1790 |

More Information
Other Species Targeted

Gene Ontology Categories:
