siRNA Details
Gene Symbol: Lsmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033437.2 | NP_001028609.1 | 4 | 747 | ||
GenBank mRNA | AK134025.1 | - | 794 | |||
AK142184.1 | - | 4 | 748 | |||
BC147386.1 | - | 4 | 667 | 9056448 | ||
BC147387.1 | - | 4 | 667 | 9056444 | ||
BC172052.1 | - | 4 | 671 |
