siRNA Details
Gene Symbol: Lsmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033437.2 | NP_001028609.1 | 4 | 1188 | ||
GenBank mRNA | AK134025.1 | - | 1234 | |||
AK142184.1 | - | 4 | 1189 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lsmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033437.2 | NP_001028609.1 | 4 | 1188 | ||
GenBank mRNA | AK134025.1 | - | 1234 | |||
AK142184.1 | - | 4 | 1189 |