siRNA Details
Gene Symbol: Gm933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256309.1 | NP_001243238.1 | 3 | 348 | ||
GenBank mRNA | AI509129.1 | - | 3 | 348 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA028844.1 | - | 348 | Chr. - - [-] on Build |
