siRNA Details
Gene Symbol: Zfp408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033451.2 | NP_001028623.1 | 4 | 1621 | ||
GenBank mRNA | AK132515.1 | - | 4 | 1622 | ||
AK134439.1 | - | 1 | 471 | |||
AK161736.1 | - | 2 | 760 | |||
BC145603.1 | - | 4 | 1245 | 9054169 | ||
BC145604.1 | - | 5 | 1393 |
