siRNA Details
Gene Symbol: Tmem240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101506.1 | NP_001094976.1 | 4 | 963 | ||
GenBank mRNA | AK147532.1 | - | 2 | 5095 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008764407, XM_008764408, XM_002729582.
Gene Ontology Categories:
