siRNA Details
Gene Symbol: Gm1673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033458.3 | NP_001028630.1 | 4 | 410 | ||
GenBank mRNA | AK131594.1 | - | 3 | 379 | ||
AK141724.1 | - | 4 | 411 | |||
BC048625.1 | - | 4 | 489 | |||
BC055794.1 | - | 4 | 361 | |||
BC100514.1 | - | 449 |
