siRNA Details
Gene Symbol: C87414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164284.1 | NP_001157756.1 | 6 | 2418 | ||
NM_001164285.1 | NP_001157757.1 | 4 | 2271 | |||
GenBank mRNA | AK139835.1 | - | 6 | 2468 | ||
AK163210.1 | - | 6 | 1910 | |||
AK163343.1 | - | 2211 | ||||
BB705621.1 | - | 1 | 208 | |||
BC052888.1 | - | 5 | 1837 | 30051439 | ||
BC080704.1 | - | 5 | 2376 | 30476802 | ||
BC083101.1 | - | 4 | 2227 | 30477931 | ||
BC085232.1 | - | 6 | 2351 | 30476183 |
Gene Symbol: Gm6367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054302.1 | - | 2253 |
