siRNA Details
Gene Symbol: C87414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164284.1 | NP_001157756.1 | 6 | 2127 | ||
NM_001164285.1 | NP_001157757.1 | 4 | 1980 | |||
GenBank mRNA | AK139835.1 | - | 6 | 2177 | ||
AK163210.1 | - | 6 | 1619 | |||
AK163343.1 | - | 1920 | ||||
BC052888.1 | - | 5 | 1546 | 30051439 | ||
BC085232.1 | - | 6 | 2060 | 30476183 |
