siRNA Details
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277450.1 | NP_001264379.1 | 2 | 302 | ||
NM_019191.2 | NP_062064.1 | 2 | 330 | |||
XM_006254945.2 | XP_006255007.1 | 2 | 416 | |||
GenBank mRNA | AB010147.1 | - | 1 | 104 | ||
AB017912.1 | - | 2 | 196 | |||
AF056001.1 | - | 2 | 230 | |||
BC127497.1 | - | 2 | 174 | 8364718 | ||
FQ230791.1 | - | 2 | 168 |
