siRNA Details
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081688.2 | NP_001075157.2 | 6 | 629 | ||
GenBank mRNA | AB109391.1 | - | 6 | 740 | ||
BC158102.1 | - | 6 | 725 | 9007387 | ||
BC172169.1 | - | 6 | 725 | 9054839 |
