siRNA Details
Gene Symbol: Gm5460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034880.2 | NP_001030052.1 | 5 | 1406 | ||
GenBank mRNA | AK142650.1 | - | 5 | 1407 | ||
BC150959.1 | - | 5 | 1370 | 9087870 | ||
BC151174.1 | - | 5 | 1370 | 9088063 |
