siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB711853.1 | - | 2, 3 | 193 | ||
CF978038.1 | - | 2, 3 | 206 | |||
M11597.1 | - | 1, 2 | 99 | |||
M26137.1 | - | 1, 2 | 106 | |||
V01228.1 | - | 2, 3 | 162 | |||
V01229.1 | - | 1, 2 | 106 | |||
V01231.1 | - | 1, 2 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
