siRNA Details
Gene Symbol: Smim22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253796.1 | NP_001240725.1 | 4 | 362 | ||
NM_001253797.1 | NP_001240726.1 | 4 | 347 | |||
NM_001253802.1 | NP_001240731.1 | 4 | 297 | |||
NM_001253803.1 | NP_001240732.1 | 4 | 282 | |||
GenBank mRNA | AW985536.1 | - | 4 | 273 | ||
BB610432.1 | - | 3 | 184 | |||
BC047216.1 | - | 4 | 341 | |||
CJ043101.1 | - | 4 | 298 | |||
CJ237462.1 | - | 4 | 344 | |||
CJ320043.1 | - | 4 | 359 |
