siRNA Details
Gene Symbol: Spink11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048217.3 | NP_001041682.2 | 4 | 229 | ||
GenBank mRNA | AK136741.1 | - | 4 | 227 | ||
BC147524.1 | - | 4 | 198 | 9056516 | ||
BC147541.1 | - | 4 | 198 | 9056532 | ||
DQ437331.1 | - | 4 | 229 |
