siRNA Details
Gene Symbol: Gm5544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033779.2 | NP_001028951.1 | 2 | 733 | ||
GenBank mRNA | AK142329.1 | - | 2 | 734 | ||
BC147112.1 | - | 1 | 567 | 9056126 | ||
BC147113.1 | - | 1 | 567 | 9056128 |
