siRNA Details
Gene Symbol: Minos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163006.2 | NP_001156478.1 | 4 | 1158 | ||
GenBank mRNA | AK030343.1 | - | 4 | 1111 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Minos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163006.2 | NP_001156478.1 | 4 | 1158 | ||
GenBank mRNA | AK030343.1 | - | 4 | 1111 |