siRNA Details
Gene Symbol: Tmem221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100462.1 | NP_001093932.1 | 4 | 1218 | ||
GenBank mRNA | BC033932.1 | - | 4 | 925 | ||
BC147816.1 | - | 4 | 1218 | 9087533 | ||
BC147831.1 | - | 4 | 1218 | 9087548 |
