siRNA Details
Gene Symbol: 1190007I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135567.1 | NP_001129039.1 | 3 | 430 | ||
NM_001135568.1 | NP_001129040.1 | 3 | 301 | |||
NM_001135569.1 | NP_001129041.1 | 3 | 242 | |||
GenBank mRNA | AK004515.1 | - | 3 | 292 | ||
AK045413.1 | - | 3 | 314 | |||
BC038652.1 | - | 3 | 260 | |||
BC065417.1 | - | 3 | 264 | |||
DV649038.1 | - | 3 | 236 |

More Information
Other Species Targeted

Gene Ontology Categories:
