siRNA Details
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019156.2 | NP_062029.2 | 3 | 444 | ||
GenBank mRNA | AY318963.1 | - | 405 | |||
BC105821.1 | - | 3 | 444 | 7371104 | ||
FQ209609.1 | - | 3 | 456 | |||
FQ210146.1 | - | 1 | 64 | |||
FQ210518.1 | - | 3 | 449 | |||
FQ212575.1 | - | 3 | 443 | |||
FQ218318.1 | - | 3 | 456 | |||
FQ218401.1 | - | 1 | 17 | |||
FQ219297.1 | - | 3 | 457 | |||
U44845.1 | - | 3 | 434 |
