siRNA Details
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033498.1 | NP_001028670.1 | 11 | 1330 | ||
GenBank mRNA | AK040545.1 | - | 12 | 1219 | ||
AK041631.1 | - | 11 | 1330 | |||
BC064463.1 | - | 2 | 2091 |
