siRNA Details
Gene Symbol: Tmem253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033805.3 | NP_001028977.1 | 7 | 910 | ||
GenBank mRNA | AK144313.1 | - | 7 | 911 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033805.3 | NP_001028977.1 | 7 | 910 | ||
GenBank mRNA | AK144313.1 | - | 7 | 911 |