siRNA Details
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160412.1 | NP_001153884.1 | 3 | 433 | ||
GenBank mRNA | AV461889.1 | - | 3 | 443 | ||
AV467090.1 | - | 1 | 79 | |||
EU599038.1 | - | 5 | 523 | |||
EU599039.1 | - | 4 | 507 | |||
EU599040.1 | - | 4 | 507 |
