siRNA Details
Gene Symbol: Zfp827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294279.1 | NP_001281208.1 | 15 | 6004 | ||
NM_178267.3 | NP_839998.2 | 15 | 5999 | |||
GenBank mRNA | AK033589.1 | - | 1 | 1856 | ||
AK035374.1 | - | 1 | 742 | |||
BC041776.1 | - | 8 | 3466 | |||
BC088739.1 | - | 10 | 3629 |
