siRNA Details
Gene Symbol: Gm6377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037917.2 | NP_001033006.1 | 2 | 992 | ||
GenBank mRNA | AK080287.1 | - | 2 | 959 | ||
AK138683.1 | - | 2 | 960 | |||
BC147431.1 | - | 2 | 956 | 9056503 | ||
BC147432.1 | - | 2 | 956 | 9056492 |
