siRNA Details
Gene Symbol: Gm14137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039223.3 | NP_001034312.1 | 1 | 2595 | ||
GenBank mRNA | AK028753.1 | - | 1 | 2596 | ||
BB011034.2 | - | 1 | 170 | |||
BC150984.1 | - | 1 | 2590 | 9087897 | ||
BC150992.1 | - | 2 | 2576 | 9087905 |
