siRNA Details
Gene Symbol: Defa26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079933.2 | NP_001073402.1 | 2 | 230 | ||
GenBank mRNA | AY766470.1 | - | 2 | 215 | ||
CJ043218.1 | - | 2 | 231 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Defa26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079933.2 | NP_001073402.1 | 2 | 230 | ||
GenBank mRNA | AY766470.1 | - | 2 | 215 | ||
CJ043218.1 | - | 2 | 231 |