siRNA Details
Gene Symbol: Klk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039042.2 | NP_001034131.2 | 4 | 542 | ||
GenBank mRNA | BC063763.1 | - | 4 | 543 |

More Information
Other Species Targeted

Gene Ontology Categories:
