siRNA Details
Gene Symbol: Gm14092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037929.2 | NP_001033018.1 | 3 | 406 | ||
GenBank mRNA | AK133124.1 | - | 3 | 407 | ||
BC147590.1 | - | 3 | 382 | 9056580 | ||
BC147611.1 | - | 3 | 382 | 9056601 |
