siRNA Details
Gene Symbol: Dcpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039238.2 | NP_001034327.2 | 4 | 335 | ||
GenBank mRNA | BC059096.1 | - | 3 | 354 | 4914089 | |
BC171996.1 | - | 336 | 9054282 | |||
DQ237937.1 | - | 335 |
