siRNA Details
Gene Symbol: Vmn2r115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104579.1 | NP_001098049.1 | 3 | 617 | ||
GenBank mRNA | AB540946.1 | - | 3 | 617 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
