siRNA Details
Gene Symbol: Gm7276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158289.1 | - | 1 | 1273 | ||
BC147247.1 | - | 1 | 1166 | 9056308 | ||
BC147248.1 | - | 1 | 1166 | 9056300 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158289.1 | - | 1 | 1273 | ||
BC147247.1 | - | 1 | 1166 | 9056308 | ||
BC147248.1 | - | 1 | 1166 | 9056300 |