siRNA Details
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019155.2 | NP_062028.1 | 2 | 290 | ||
GenBank mRNA | BC084692.1 | - | 2 | 290 | 7103841 | |
U31968.1 | - | 2 | 288 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019155.2 | NP_062028.1 | 2 | 290 | ||
GenBank mRNA | BC084692.1 | - | 2 | 290 | 7103841 | |
U31968.1 | - | 2 | 288 |