siRNA Details
Gene Symbol: Tmem243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081029.1 | NP_001074498.1 | 5 | 860 | ||
GenBank mRNA | AK076719.1 | - | 4 | 720 |

More Information
Other Species Targeted

Gene Ontology Categories:
