siRNA Details
Gene Symbol: Gm7337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133184.1 | - | 1 | 1127 | ||
BC147596.1 | - | 1 | 1050 | 9056586 | ||
BC147623.1 | - | 1 | 1050 | 9056613 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133184.1 | - | 1 | 1127 | ||
BC147596.1 | - | 1 | 1050 | 9056586 | ||
BC147623.1 | - | 1 | 1050 | 9056613 |