siRNA Details
Gene Symbol: Smco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039558.2 | NP_001034647.2 | 1, 2 | 99 | ||
GenBank mRNA | AK045581.1 | - | 1, 2 | 76 | ||
AK078719.1 | - | 1, 2 | 102 | |||
BB624054.1 | - | 1, 2 | 99 |
