siRNA Details
Gene Symbol: Cxx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024170.2 | NP_077132.1 | 1 | 994 | ||
GenBank mRNA | AK002323.1 | - | 1 | 929 | ||
BC003436.1 | - | 1 | 913 | 2647372 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024170.2 | NP_077132.1 | 1 | 994 | ||
GenBank mRNA | AK002323.1 | - | 1 | 929 | ||
BC003436.1 | - | 1 | 913 | 2647372 |