siRNA Details
Gene Symbol: Commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033132.3 | NP_001028304.1 | 4 | 575 | ||
NM_001168592.1 | NP_001162064.1 | 3 | 563 | |||
GenBank mRNA | AK004316.1 | - | 4 | 549 | ||
AK020234.1 | - | 4 | 548 | |||
AK045407.1 | - | 3049 | ||||
CK128085.1 | - | 1 | 326 | |||
CK621880.1 | - | 3 | 526 |
