siRNA Details
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019167.1 | NP_062040.1 | 16, 17 | 3972 | ||
XM_008760092.1 | XP_008758314.1 | 15, 16 | 4204 | |||
XM_008760093.1 | XP_008758315.1 | 15, 16 | 4204 | |||
XM_008760094.1 | XP_008758316.1 | 16, 17 | 3701 | |||
GenBank mRNA | AB001347.1 | - | 16, 17 | 3972 |
