siRNA Details
Gene Symbol: Gm7534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080712.1 | NP_001074181.1 | 1 | 897 | ||
GenBank mRNA | AK138880.1 | - | 1 | 897 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080712.1 | NP_001074181.1 | 1 | 897 | ||
GenBank mRNA | AK138880.1 | - | 1 | 897 |