siRNA Details
Gene Symbol: Rbm24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081425.1 | NP_001074894.1 | 1 | 762 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191100.
Gene Ontology Categories:
