siRNA Details
Gene Symbol: BC023105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023105.1 | - | 2 | 247 | 3989161 | |
BF163606.1 | - | 2 | 247 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC023105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023105.1 | - | 2 | 247 | 3989161 | |
BF163606.1 | - | 2 | 247 |