siRNA Details
Gene Symbol: Gm8787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099310.1 | NP_001092780.1 | 3, 4 | 469 | ||
GenBank mRNA | BY735733.1 | - | 3, 4 | 469 |

More Information
Other Species Targeted

Gene Ontology Categories:
