siRNA Details
Gene Symbol: 4930453N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026273.2 | NP_080549.1 | 3 | 1009 | ||
GenBank mRNA | AK002578.1 | - | 3 | 1004 | ||
AK075826.1 | - | 3 | 993 | |||
AK161311.1 | - | 3 | 1010 | |||
BC020029.1 | - | 3 | 998 | 4012943 |
