siRNA Details
Gene Symbol: 4930523C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 2478 | ||
GenBank mRNA | AK028660.1 | - | 517 | |||
AK028922.1 | - | 3 | 2281 | |||
AK034159.1 | - | 3 | 2173 | |||
AK054156.1 | - | 75 |
