siRNA Details
Gene Symbol: Klk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026806.2 | NP_081082.1 | 3 | 423 | ||
GenBank mRNA | AK003996.1 | - | 3 | 424 | ||
BC139475.1 | - | 3 | 424 | 8862497 | ||
BC139478.1 | - | 3 | 424 | 8862500 |
