siRNA Details
Gene Symbol: Fam172a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163419.1 | NP_001156891.1 | 10 | 3276 | ||
NM_001163420.1 | NP_001156892.1 | 9 | 3084 | |||
NM_138312.1 | NP_612185.1 | 11 | 3400 | |||
GenBank mRNA | AK052419.1 | - | 10 | 3249 | ||
BC057380.1 | - | 10 | 3244 |
