siRNA Details
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027009.2 | NP_081285.1 | 9 | 1161 | ||
GenBank mRNA | AK145126.1 | - | 9 | 1188 | ||
AK165891.1 | - | 6 | 2672 | |||
AK167047.1 | - | 9 | 1162 | |||
BC026795.1 | - | 9 | 1132 | 4015134 |
