siRNA Details
Gene Symbol: 1700008O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027049.1 | NP_081325.1 | 6 | 884 | ||
GenBank mRNA | AK005784.1 | - | 6 | 884 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106255, XM_006228989, XM_008759332.
Gene Ontology Categories:
