siRNA Details
Gene Symbol: 1700019A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027070.1 | NP_081346.1 | 5, 6 | 304 | ||
GenBank mRNA | AK006101.1 | - | 5, 6 | 305 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700019A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027070.1 | NP_081346.1 | 5, 6 | 304 | ||
GenBank mRNA | AK006101.1 | - | 5, 6 | 305 |